1-(Methylsulfonyl)piperidine-4-carbaldehyde ≥97%

Supplier: Matrix Scientific

081729-1G
102835-666EA 949.23 USD
102835-666
1-(Methylsulfonyl)piperidine-4-carbaldehyde ≥97%
1-(Methylsulfonyl)piperidine-4-carbaldehyde

Hazard: Irritant

Formula: C₇H₁₃NO₃S
MW: 191.25 g/mol
MDL Number: MFCD11582937
CAS Number: 241134-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR