2-Chloroquinoline-3-carbaldehyde ≥97%

Supplier: Matrix Scientific

081718-1G 081718-10G
102835-636EA 127.69 USD
102835-636 102835-638
2-Chloroquinoline-3-carbaldehyde ≥97%
2-Chloroquinoline-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₆ClNO
MW: 191.62 g/mol
MDL Number: MFCD00130079
CAS Number: 73568-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR