3-Quinolinecarbaldehyde ≥97%
Supplier: Matrix Scientific
Synonyms:
Quinoline-3-carbaldehyde
, Quinoline-3-carboxaldehyde
, 3-Quinolinecarboxaldehyde
081716-1G
102835-628EA
46.15
USD
102835-628
3-Quinolinecarbaldehyde ≥97%
3-Quinolinecarbaldehyde
Hazard: Irritant
Formula:
C₁₀H₇NO MW: 157.17 g/mol Melting Pt: 68…71 °C |
MDL Number:
MFCD00006768 CAS Number: 13669-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...