1-Methylimidazole-4-carbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Methyl-1H-imidazole-4-carbaldehyde
081704-1G
102835-600EA
1287.69
USD
102835-600
1-Methylimidazole-4-carbaldehyde ≥95%
1-Methylimidazole-4-carbaldehyde
Hazard: Irritant
Formula:
C₅H₆N₂O MW: 110.1139 g/mol |
MDL Number:
MFCD03411957 CAS Number: 17289-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...