1-Tritylimidazole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
081701-1G
102835-590EA
353.85
USD
102835-590
1-Tritylimidazole-2-carbaldehyde ≥95%
1-Tritylimidazole-2-carbaldehyde
Hazard: Irritant
Formula:
C₂₃H₁₈N₂O |
MDL Number:
MFCD03453110 CAS Number: 67478-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...