2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-chloro-3-cyano-5-formylthiophene2-Amino-4-chloro-5-formyl-3-thiophenecarbonitrile

081657-1G
102835-488EA 383.08 USD
102835-488
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥97%
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile

Hazard: Irritant

Formula: C₆H₃ClN₂OS
MW: 186.62 g/mol
MDL Number: MFCD03788265
CAS Number: 104366-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR