6-(4-Fluorophenyl)-2-pyridinecarbaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde

081643-1G
102835-456EA 384.62 USD
102835-456
6-(4-Fluorophenyl)-2-pyridinecarbaldehyde ≥97%
6-(4-Fluorophenyl)-2-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₂H₈FNO
MDL Number: MFCD06410011
CAS Number: 834884-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR