4-Formyl-3-picoline ≥95%

Supplier: Matrix Scientific

081626-1G
102835-420EA 563.5 USD
102835-420
4-Formyl-3-picoline ≥95%
4-Formyl-3-picoline

Hazard: Irritant

Formula: C₇H₇NO
MDL Number: MFCD07437997
CAS Number: 51227-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR