(4-Bromo-2-fluorophenyl)acetaldehyde ≥95%
Supplier: Matrix Scientific
081613-1G
102835-394EA
863.08
USD
102835-394
(4-Bromo-2-fluorophenyl)acetaldehyde ≥95%
(4-Bromo-2-fluorophenyl)acetaldehyde
Hazard: Irritant
Formula:
C₈H₆BrFO |
MDL Number:
MFCD09991763 CAS Number: 916516-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...