(4-Methylphenyl)acetaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(p-Tolyl)acetaldehyde

081611-1G
102835-390EA 361.54 USD
102835-390
(4-Methylphenyl)acetaldehyde ≥95%
(4-Methylphenyl)acetaldehyde

Hazard: Irritant

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00047658
CAS Number: 104-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR