4-Benzyloxy-2-methoxybenzaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Benzyloxy)-2-methoxybenzaldehyde

081596-1G
102835-360EA 127.69 USD
102835-360
4-Benzyloxy-2-methoxybenzaldehyde ≥97%
4-Benzyloxy-2-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MW: 242.2700 g/mol
MDL Number: MFCD01075699
CAS Number: 58026-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR