2,3-Dichloro-4-hydroxybenzaldehyde ≥97%

Supplier: Matrix Scientific

081585-1G
102835-326EA 423.2 USD
102835-326
2,3-Dichloro-4-hydroxybenzaldehyde ≥97%
2,3-Dichloro-4-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₇H₄Cl₂O₂
MDL Number: MFCD07775189
CAS Number: 16861-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR