3,5-Difluoro-4-hydroxybenzaldehyde ≥97%

Supplier: Matrix Scientific

081584-1G
102835-324EA 609.23 USD
102835-324
3,5-Difluoro-4-hydroxybenzaldehyde ≥97%
3,5-Difluoro-4-hydroxybenzaldehyde

Hazard: Irritant

Formula: C₇H₄F₂O₂
MDL Number: MFCD06797917
CAS Number: 118276-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR