4-(Pentafluorothio)benzaldehyde ≥99%

Supplier: Matrix Scientific

081582-1G
102835-320EA 767.69 USD
102835-320
4-(Pentafluorothio)benzaldehyde ≥99%
4-(Pentafluorothio)benzaldehyde

Hazard: Irritant

Formula: C₇H₅F₅OS
MDL Number: MFCD03788518
CAS Number: 401892-84-0
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR