3-(Pyrimidin-2-yl)benzaldehyde ≥95%

Supplier: Matrix Scientific

081562-1G
102835-276EA 566.15 USD
102835-276
3-(Pyrimidin-2-yl)benzaldehyde ≥95%
3-(Pyrimidin-2-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₁H₈N₂O
MDL Number: MFCD08056283
CAS Number: 263349-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR