3-Fluoro-4-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: Matrix Scientific

081545-1G
102835-230EA 241.54 USD
102835-230
3-Fluoro-4-(trifluoromethoxy)benzaldehyde ≥97%
3-Fluoro-4-(trifluoromethoxy)benzaldehyde

Hazard: Irritant

Formula: C₈H₄F₄O₂
MDL Number: MFCD00236275
CAS Number: 473917-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR