2-Chloro-4-(dimethylamino)benzaldehyde ≥97%

Supplier: Matrix Scientific

081541-1G 081541-10G
102835-220EA 255.38 USD
102835-220 102835-222
2-Chloro-4-(dimethylamino)benzaldehyde ≥97%
2-Chloro-4-(dimethylamino)benzaldehyde

Hazard: Irritant

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
MDL Number: MFCD00021051
CAS Number: 1424-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR