4-Methylsalicylaldehyde ≥97%
Synonyms:
2-Hydroxy-4-methylbenzaldehyde
081539-1G
081539-10G
102835-216EA
861.1
USD
102835-216
102835-214
4-Methylsalicylaldehyde ≥97%
4-Methylsalicylaldehyde
Hazard: Irritant
Formula:
C₈H₈O₂ MW: 136.15 g/mol |
MDL Number:
MFCD00799550 CAS Number: 698-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...