Dipropylamine hydrochloride ≥97%
081516-1G
081516-10G
081516-50G
102835-140EA
861.11
USD
102835-140
102835-144
102835-142
Dipropylamine hydrochloride ≥97%
Dipropylamine hydrochloride
Purity: 97%
CAS: 5326-84-1
MDL Number: MFCD00054288
Molecular Formula: C6H16ClN
Molecular Weight: 137,65
Hazard: Irritant
Formula:
C₆H₁₅N |
MDL Number:
MFCD00054288 CAS Number: 5326-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...