5-Amino-3-bromo-1H-indazole ≥97%

Supplier: Matrix Scientific

081498-1G
102835-082EA 356.5 USD
102835-082
5-Amino-3-bromo-1H-indazole ≥97%
5-Amino-3-bromo-1H-indazole

Hazard: Irritant

Formula: C₇H₆BrN₃
MDL Number: MFCD07781655
CAS Number: 478837-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR