2-Aminopurine ≥97%

Supplier: Matrix Scientific

Synonyms: 7H-Purin-2-aminePurin-2-amine

081489-1G
102835-058EA 198.46 USD
102835-058
2-Aminopurine ≥97%
2-Aminopurine

Hazard: Irritant

Formula: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: 278 °C
MDL Number: MFCD00005566
CAS Number: 452-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR