3-Amino-4-chloro-1-methyl-1H-indazole ≥97%
Supplier: Matrix Scientific
081480-1G
102835-036EA
636.92
USD
102835-036
3-Amino-4-chloro-1-methyl-1H-indazole ≥97%
3-Amino-4-chloro-1-methyl-1H-indazole
Hazard: Irritant
Formula:
C₈H₈ClN₃ |
MDL Number:
MFCD09864571 CAS Number: 959240-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...