2-Amino-6-iodopurine ≥97%

Supplier: Matrix Scientific

081457-1G
102834-984EA 707.69 USD
102834-984
2-Amino-6-iodopurine ≥97%
2-Amino-6-iodopurine

Hazard: Irritant

Formula: C₅H₄IN₅
MW: 261.02
MDL Number: MFCD08706375
CAS Number: 19690-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR