3-Aminocoumarin ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2H-chromen-2-one

081453-1G
102834-972EA 750.77 USD
102834-972
3-Aminocoumarin ≥97%
3-Aminocoumarin

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00016965
CAS Number: 1635-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR