1-Aminobenzotriazole ≥97%
Supplier: Matrix Scientific
Synonyms:
1H-Benzo[d][1,2,3]triazol-1-amine
081452-1G
102834-970EA
496.92
USD
102834-970
1-Aminobenzotriazole ≥97%
1-Aminobenzotriazole
Hazard: Irritant
Formula:
C₆H₆N₄ MW: 134.14 g/mol |
MDL Number:
MFCD00132902 CAS Number: 1614-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...