(S)-(-)-3-Aminoquinuclidine dihydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
(S)-Quinuclidin-3-amine dihydrochloride
, (S)-(-)-3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride
081438-50G
081438-1G
081438-10G
102834-936EA
3326.17
USD
102834-936
102834-932
102834-934
(S)-(-)-3-Aminoquinuclidine dihydrochloride ≥97%
(S)-(-)-3-Aminoquinuclidine dihydrochloride
Hazard: Irritant
Formula:
C₇H₁₆Cl₂N₂ MW: 199.1213 g/mol |
MDL Number:
MFCD00191753 CAS Number: 119904-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...