2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Methoxy-N,6-dimethyl-1,3,5-triazin-2-amine
081429-10G
081429-50G
081429-1G
102834-902EA
473.85
USD
102834-902
102834-898
102834-900
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine ≥97%
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine
Hazard: Irritant
Formula:
C₆H₁₀N₄O MW: 154.17 g/mol |
MDL Number:
MFCD00585858 CAS Number: 5248-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...