1-Boc-3-(methylamino)piperidine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Methylamino)piperidine, N1-BOC protected1-Boc-3-methylaminopieridine

081398-1G
102834-800EA 264.5 USD
102834-800
1-Boc-3-(methylamino)piperidine ≥97%
1-Boc-3-(methylamino)piperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD04114980
CAS Number: 392331-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR