4-Chloro-2,6-diaminopyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Chloropyrimidine-2,4-diamine2,4-Diamino-6-chloropyrimidine

081364-50G 081364-10G 081364-1G
102834-712EA 116.92 USD
102834-712 102834-714 102834-710
4-Chloro-2,6-diaminopyrimidine ≥97%
4-Chloro-2,6-diaminopyrimidine

Hazard: Irritant

Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD00006097
CAS Number: 156-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR