2-Morpholin-4-ylpyrimidin-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-(morpholin-4-yl)pyrimidine

081356-1G
102834-692EA 720 USD
102834-692
2-Morpholin-4-ylpyrimidin-5-amine ≥95%
2-Morpholin-4-ylpyrimidin-5-amine

Hazard: Irritant

Formula: C₈H₁₂N₄O
MW: 180.21 g/mol
MDL Number: MFCD10703512
CAS Number: 65735-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR