2-(Methylthio)pyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-(methylthio)pyrimidine

081352-1G
102834-682EA 410.77 USD
102834-682
2-(Methylthio)pyrimidin-4-amine ≥97%
2-(Methylthio)pyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₇N₃S
MW: 141.1942 g/mol
MDL Number: MFCD02091098
CAS Number: 2183-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR