3-Amino-4-bromo-5-methylisoxazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-bromo-5-methylisoxazol-3-amine

081331-1G
102834-628EA 170.77 USD
102834-628
3-Amino-4-bromo-5-methylisoxazole ≥97%
3-Amino-4-bromo-5-methylisoxazole

Hazard: Irritant

Formula: C₄H₅BrN₂O
MW: 177.02 g/mol
MDL Number: MFCD00052553
CAS Number: 5819-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR