5-Amino-2-chloro-3-methylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-5-methylpyridin-3-amine5-Amino-2-chloro-3-picoline

081311-1G
102834-578EA 15.38 USD
102834-578
5-Amino-2-chloro-3-methylpyridine ≥97%
5-Amino-2-chloro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD03095087
CAS Number: 38186-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR