3-Amino-6-chloropyridine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

081305-1G
102834-564EA 666.15 USD
102834-564
3-Amino-6-chloropyridine-2-carbonitrile ≥97%
3-Amino-6-chloropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₄ClN₃
MDL Number: MFCD07780767
CAS Number: 95095-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR