3-Amino-4-ethoxypyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Ethoxypyridin-3-amine

081303-1G
102834-560EA 723.08 USD
102834-560
3-Amino-4-ethoxypyridine ≥97%
3-Amino-4-ethoxypyridine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00234351
CAS Number: 1633-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR