2,6-Diamino-3,5-difluoropyridine ≥97%

Supplier: Matrix Scientific

081298-1G 081298-10G
102834-548EA 353.85 USD
102834-548 102834-550
2,6-Diamino-3,5-difluoropyridine ≥97%
2,6-Diamino-3,5-difluoropyridine

Hazard: Irritant

Formula: C₅H₅F₂N₃
MW: 145.11 g/mol
MDL Number: MFCD03412208
CAS Number: 247069-27-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR