1-(3-Aminopyridin-4-yl)ethanone ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Acetyl-3-aminopyridine
081291-1G
102834-534EA
693.85
USD
102834-534
1-(3-Aminopyridin-4-yl)ethanone ≥98%
1-(3-Aminopyridin-4-yl)ethanone
Hazard: Irritant
Formula:
C₇H₈N₂O MW: 136.15 g/mol |
MDL Number:
MFCD07375078 CAS Number: 13210-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...