3-Amino-5-methylpyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

081264-1G
102834-468EA 29.23 USD
102834-468
3-Amino-5-methylpyridin-2(1H)-one ≥95%
3-Amino-5-methylpyridin-2(1H)-one

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
MDL Number: MFCD09839282
CAS Number: 52334-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR