3-Amino-4,6-dimethylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 4,6-Dimethylpyridin-3-amine

081259-1G
102834-458EA 120 USD
102834-458
3-Amino-4,6-dimethylpyridine ≥97%
3-Amino-4,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₁₀N₂
MW: 122.1677 g/mol
MDL Number: MFCD07439969
CAS Number: 1193-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR