3-Amino-2,4-dimethylpyridine ≥97%

Supplier: Matrix Scientific

081256-1G
102834-450EA 298.46 USD
102834-450
3-Amino-2,4-dimethylpyridine ≥97%
3-Amino-2,4-dimethylpyridine

Hazard: Irritant

Formula: C₇H₁₀N₂
MDL Number: MFCD08235192
CAS Number: 1073-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR