tert-Butyl-2-chloropyridin-3-ylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Chloro-pyridin-3-yl)-carbamic acid tert-butyl ester2-Methyl-2-propanyl (2-chloro-3-pyridinyl)carbamate

081254-1G
102834-444EA 750.77 USD
102834-444
tert-Butyl-2-chloropyridin-3-ylcarbamate ≥97%
tert-Butyl-2-chloropyridin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD12026344
CAS Number: 209798-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR