2-(Boc-amino)-6-chloropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-chloropyridine, 2-BOC protectedtert-Butyl (6-chloropyridin-2-yl)carbamate

081252-1G
102834-440EA 166.15 USD
102834-440
2-(Boc-amino)-6-chloropyridine ≥97%
2-(Boc-amino)-6-chloropyridine

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD09030821
CAS Number: 159603-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR