4-Morpholino-2-pyridinamine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Morpholinopyridin-2-amine2-Amino-4-(morpholin-4-yl)pyridine

081218-1G
102834-360EA 526.15 USD
102834-360
4-Morpholino-2-pyridinamine ≥97%
4-Morpholino-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃N₃O
MW: 179.2200 g/mol
MDL Number: MFCD09991750
CAS Number: 722549-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR