3-Amino-4-hydroxybenzonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-cyanophenol

081213-1G
102834-350EA 580 USD
102834-350
3-Amino-4-hydroxybenzonitrile ≥97%
3-Amino-4-hydroxybenzonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD00234273
CAS Number: 14543-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR