4-(Trimethylsilylethynyl)aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-((Trimethylsilyl)ethynyl)aniline4-[(Trimethylsilyl)ethynyl]aniline

081182-1G
102834-276EA 524.62 USD
102834-276
4-(Trimethylsilylethynyl)aniline ≥97%
4-(Trimethylsilylethynyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₅NSi
MW: 189.33 g/mol
MDL Number: MFCD03695479
CAS Number: 75867-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR