2,4,6-Tris(trifluoromethyl)aniline ≥97%

Supplier: Matrix Scientific

081176-1G
102834-260EA 1273.85 USD
102834-260
2,4,6-Tris(trifluoromethyl)aniline ≥97%
2,4,6-Tris(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₉H₄F₉N
MW: 297.12 g/mol
CAS Number: 25753-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR