2-Chloroanilinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloroaniline hydrochloride

081172-50G 081172-10G 081172-1G
102834-250EA 78.46 USD
102834-250 102834-248 102834-252
2-Chloroanilinium chloride ≥97%
2-Chloroanilinium chloride

Hazard: Irritant

Formula: C₆H₇Cl₂N
MDL Number: MFCD00053282
CAS Number: 137-04-2
UN: 2018
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR