4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4'-chloro-5-fluorobiphenyl

081136-1G
102834-140EA 636.92 USD
102834-140
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine ≥97%
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine

Hazard: Irritant

Formula: C₁₂H₉ClFN
MW: 221.66 g/mol
MDL Number: MFCD12824033
CAS Number: 188731-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR