tert-Butyl-3-chloro-2-methylphenylcarbamate ≥97%

Supplier: Matrix Scientific

081135-1G
102834-138EA 553.85 USD
102834-138
tert-Butyl-3-chloro-2-methylphenylcarbamate ≥97%
tert-Butyl-3-chloro-2-methylphenylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₂
MW: 241.72 g/mol
MDL Number: MFCD07127715
CAS Number: 129822-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR