4-(Pentafluorothio)aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Aminophenylsulphur pentafluoride

081132-1G
102834-132EA 383.08 USD
102834-132
4-(Pentafluorothio)aniline ≥97%
4-(Pentafluorothio)aniline

Hazard: Irritant

Formula: C₆H₆F₅NS
MW: 219.18
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR